Xylylazo violet I

Supplier: TCI America

A5007-1G A5007-5G
TCA5007-001GEA 86.9 CAD
TCA5007-001G TCA5007-5G
Xylylazo violet I
Xylylazo Violet I

[Spectrophotometric reagent for Mg]
CAS Number: 14936-97-1
MDL Number: MFCD00003890
Molecular Formula: C25H21N3O6S
Molecular Weight: 513.50
Form: Crystal
Color: Deep Yellow Red


Other Product Names / Synonyms: 1-Azo-2-hydroxy-3-(2,4-dimethylcarboxyanilido)naphthalene-1'-(2-hydroxybenzene-5-sulfonic Acid) Sodium Salt, 2-[2-Hydroxy-3-(2,4-xylylcarbamoyl)-1-naphthylazo]-1-phenol-4-sulfonic Acid Sodium Salt, Xylidyl Blue I

Formula: C₂₅H₂₁N₃O₆S
MDL Number: MFCD00003890
CAS Number: 14936-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fe8b0c1a8f52d68a746002bf92a5a43152ef043&vid=4fe8b0c1a8f52d68a746002bf92a5a43152ef043&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR