1,3,4,6-Tetrathiapentalene-2,5-dione ≥98.0% (by HPLC)
Supplier: TCI America
T1132-1G
T1132-5G
T1132-25G
TCT1132-001GEA
181.7
CAD
TCT1132-001G
TCT1132-005G
TCT1132-25G
1,3,4,6-Tetrathiapentalene-2,5-dione ≥98.0% (by HPLC)
1,3,4,6-Tetrathiapentalene-2,5-dione
CAS Number: 64394-45-2
MDL Number: MFCD00010403
Molecular Formula: C4O2S4
Molecular Weight: 208.28
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 178
Formula:
C₄O₂S₄ Melting Pt: 178 °C |
MDL Number:
MFCD00010403 CAS Number: 64394-45-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...