Xylylazo violet II ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: Xylidyl Blue II, 1-Azo-2-hydroxy-3-(2,4-dimethylcarboxyanilido)naphthalene-1-(2-hydroxybenzene), 2-[2-Hydroxy-3-(2,4-xylylcarbamoyl)-1-naphthylazo]phenol

A5006-1G A5006-5G
TCA5006-001GEA 319.16 CAD
TCA5006-001G TCA5006-5G
Xylylazo violet II ≥95.0% (by HPLC)
Xylylazo violet II

[Spectrophotometric reagent for Mg]
CAS Number: 523-67-1
MDL Number: MFCD00014309
Molecular Formula: C25H21N3O3
Molecular Weight: 411.46
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 247


Product Categories: Analytical Science, Azo Compounds (Chelating Agents)
Other Product Names / Synonyms: 1-Azo-2-hydroxy-3-(2,4-dimethylcarboxyanilido)naphthalene-1-(2-hydroxybenzene), 2-[2-Hydroxy-3-(2,4-xylylcarbamoyl)-1-naphthylazo]phenol, Xylidyl Blue II

Formula: C₂₅H₂₁N₃O₃
MW: 411.46 g/mol
Melting Pt: 247 °C
MDL Number: MFCD00014309
CAS Number: 523-67-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c111ddee0f25d8bdf6cf4d13ac6cf6852853054&vid=4c111ddee0f25d8bdf6cf4d13ac6cf6852853054&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR