(±)-1,2,3,4-Tetrahydro-1-naphthol ≥95.0%

Supplier: TCI America

T1111-25G T1111-5G
TCT1111-025GEA 334.39 CAD
TCT1111-025G TCT1111-5G
(±)-1,2,3,4-Tetrahydro-1-naphthol ≥95.0%
(±)-1,2,3,4-Tetrahydro-1-naphthol

CAS Number: 529-33-9
MDL Number: MFCD00001739
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >95.0% (GC)
Form: Lump
Color: White
Boiling point (°C): 104
Flash Point (°C): 113
Freezing point (°C): 28


Other Product Names / Synonyms: ac-alpha-Tetralol

Formula: C₁₀H₁₂O
Boiling Pt: 104 °C
Flash Pt: 113 °C
MDL Number: MFCD00001739
CAS Number: 529-33-9
EINECS: 208-459-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8e19a9b044752487db9a990521917286cc8cc0f&vid=e8e19a9b044752487db9a990521917286cc8cc0f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR