3,4,5-Trimethoxybenzoic anhydride 95,0

Supplier: TCI America

T1093-25G T1093-5G
TCT1093-025GEA 556.16 CAD
TCT1093-025G TCT1093-005G
3,4,5-Trimethoxybenzoic anhydride 95,0
3,4,5-Trimethoxybenzoic anhydride

CAS Number: 1719-88-6
MDL Number: MFCD00017155
Molecular Formula: C20H22O9
Molecular Weight: 406.39
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 159

Formula: C₂₀H₂₂O₉
MW: 406.383 g/mol
Melting Pt: 159 °C
MDL Number: MFCD00017155
CAS Number: 1719-88-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a164d180281f516daf5d4ecc0f68357ed702747c&vid=a164d180281f516daf5d4ecc0f68357ed702747c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR