2,2'-Dihydroxyazobenzene ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A5005-1G A5005-5G
TCA5005-001GEA 213.84 CAD
TCA5005-001G TCA5005-5G
2,2'-Dihydroxyazobenzene ≥98.0% (by HPLC, titration analysis)
2,2'-Dihydroxyazobenzene

[Spectrophotometric and fluorimetric reagent for Al, Mg and other metals]
CAS Number: 2050-14-8
MDL Number: MFCD00002182
Molecular Formula: C12H10N2O2
Molecular Weight: 214.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 175


Product Categories: Analytical Science, Azo Compounds (Chelating Agents)
Other Product Names / Synonyms: 2,2'-Azodiphenol, DHAB

Formula: C₁₂H₁₀N₂O₂
Melting Pt: 175 °C
MDL Number: MFCD00002182
CAS Number: 2050-14-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e40aaa6738294043c9b07ad6a011fdfbcb0ad0a5&vid=e40aaa6738294043c9b07ad6a011fdfbcb0ad0a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR