Potassium monomethyl terephthalate ≥95.0%

Supplier: TCI America

Synonyms: Terephthalic Acid Monomethyl Ester Potassium Salt

T1083-25G T1083-500G
TCT1083-025GEA 47.4 CAD
TCT1083-025G TCT1083-500G
Potassium monomethyl terephthalate ≥95.0%
Potassium monomethyl terephthalate

CAS Number: 42967-55-5
MDL Number: MFCD00058989
Molecular Formula: C9H8O4
Molecular Weight: 218.25
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Terephthalic Acid Monomethyl Ester Potassium Salt

Formula: C₉H₈O₄
MDL Number: MFCD00058989
CAS Number: 42967-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d490f797c9e2e3bb0d5acb8096c2d5f22c0941b&vid=6d490f797c9e2e3bb0d5acb8096c2d5f22c0941b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR