Di-p-tolyldisulfide ≥98.0%

Supplier: TCI America

Synonyms: p-Tolyl disulfide, p-Tolyl disulphide, p-Tolyl Disulfide, p-Tolyl Disulphide

T1074-25G T1074-100G
TCT1074-025GEA 163.69 CAD
TCT1074-025G TCT1074-100G
Di-p-tolyldisulfide ≥98.0%
Di-p-tolyldisulfide

CAS Number: 103-19-5
MDL Number: MFCD00008547
Molecular Formula: C14H14S2
Molecular Weight: 246.39
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 45
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Sulfenylation
Other Product Names / Synonyms: p-Tolyl Disulfide

Formula: C₁₄H₁₄S₂
Melting Pt: 45 °C
Flash Pt: 110 °C
MDL Number: MFCD00008547
CAS Number: 103-19-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b446a4a1d99ed7c97fabf6c5969ffd77e3bddd7d&vid=b446a4a1d99ed7c97fabf6c5969ffd77e3bddd7d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR