4,4',4''-Tris(benzoyloxy)trityl bromide ≥97.0%

Supplier: TCI America

Synonyms: Tris[4-(phenylcarbonyloxy)phenyl]methyl Bromide

T1071-25G T1071-5G
TCT1071-25GEA 2194.62 CAD
TCT1071-25G TCT1071-005G
4,4',4''-Tris(benzoyloxy)trityl bromide ≥97.0%
4,4',4''-Tris(benzoyloxy)trityl bromide

[Hydroxyl Protecting Agent]
CAS Number: 86610-66-4
MDL Number: MFCD00010302
Molecular Formula: C40H27BrO6
Molecular Weight: 683.55
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 190


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Nucleic Acid Synthesis
Other Product Names / Synonyms: Tris[4-(phenylcarbonyloxy)phenyl]methyl Bromide

Formula: C₄₀H₂₇BrO₆
MW: 683.55 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00010302
CAS Number: 86610-66-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59b0b74a46d1a7da3f2dfb4cd5e166496cd762ff&vid=59b0b74a46d1a7da3f2dfb4cd5e166496cd762ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR