2,2'-Thiodiethylenebis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥98.0%
Supplier: TCI America
Synonyms:
2,2-Thiodiethylene Bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]
T1070-500G
T1070-25G
T1070-100G
TCT1070-500GEA
519.82
CAD
TCT1070-500G
TCT1070-025G
TCT1070-100G
2,2'-Thiodiethylenebis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥98.0%
2,2'-Thiodiethylenebis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]
CAS Number: 41484-35-9
MDL Number: MFCD00059346
Molecular Formula: C38H58O6S
Molecular Weight: 642.94
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 78
Formula:
C₃₈H₅₈O₆S MW: 642.94 g/mol Melting Pt: 78 °C |
MDL Number:
MFCD00059346 CAS Number: 41484-35-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...