3,3',5,5'-Tetra-tert-butyl-4,4'-stilbenequinone ≥95.0%

Supplier: TCI America

T1067-5G T1067-1G
TCT1067-5GEA 1173.94 CAD
TCT1067-5G TCT1067-001G
3,3',5,5'-Tetra-tert-butyl-4,4'-stilbenequinone ≥95.0%
3,3',5,5'-Tetra-tert-butyl-4,4'-stilbenequinone

CAS Number: 809-73-4
MDL Number: MFCD00060073
Molecular Formula: C30H42O2
Molecular Weight: 434.66
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: Yellow

Formula: C₃₀H₄₂O₂
MDL Number: MFCD00060073
CAS Number: 809-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44b4278a59877a090bbaaa43fc74e1674435dea7&vid=44b4278a59877a090bbaaa43fc74e1674435dea7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR