8-Mercaptoquinoline hydrochloride 95,0
Supplier: TCI America
Synonyms:
Thiooxine Hydrochloride, 8-Mercaptoquinoline Hydrochloride
A5004-5G
A5004-1G
TCA5004-5GEA
742.6
CAD
TCA5004-5G
TCA5004-001G
8-Mercaptoquinoline hydrochloride 95,0
8-Mercaptoquinoline hydrochloride
[Extraction-spectrophotometric and fluorimetric reagent for soft metals]
CAS Number: 34006-16-1
MDL Number: MFCD00043261
Molecular Formula: C9H7NS
Molecular Weight: 197.68
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Product Categories: Analytical Science, Bipyridyls, etc. (Chelating Agents)
Other Product Names / Synonyms: Thiooxine Hydrochloride
Formula:
C₉H₇NS MW: 197.68 g/mol |
MDL Number:
MFCD00043261 CAS Number: 34006-16-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...