1,1,3,3-Tetrabromoacetone ≥98.0%

Supplier: TCI America

Synonyms: 1,1,3,3-Tetrabromo-2-propanone

T1026-5G T1026-25G
TCT1026-005GEA 48.98 CAD
TCT1026-005G TCT1026-25G
1,1,3,3-Tetrabromoacetone ≥98.0%
1,1,3,3-Tetrabromoacetone

CAS Number: 22612-89-1
MDL Number: MFCD00059470
Molecular Formula: C3H2Br4O
Molecular Weight: 373.66
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 38
Storage Temperature: <0°C


Other Product Names / Synonyms: 1,1,3,3-Tetrabromo-2-propanone

Formula: C₃H₂Br₄O
Melting Pt: 38 °C
MDL Number: MFCD00059470
CAS Number: 22612-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24c4ce067774407ba1689d4c2520babda4889b60&vid=24c4ce067774407ba1689d4c2520babda4889b60&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR