3-Amino-2,6-dibromopyridine ≥98.0%

Supplier: TCI America

Synonyms: 2,6-Dibromopyridin-3-amine

A2272-1G A2272-5G
TCA2272-1GEA 60.04 CAD
TCA2272-1G TCA2272-5G
3-Amino-2,6-dibromopyridine ≥98.0%
3-Amino-2,6-dibromopyridine

CAS Number: 39856-57-0
MDL Number: MFCD00128862
Molecular Formula: C5H4Br2N2
Molecular Weight: 251.91
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 147

Formula: C₅H₄Br₂N₂
MW: 251.9100 g/mol
Melting Pt: 147 °C
MDL Number: MFCD00128862
CAS Number: 39856-57-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b46aa5d466a159b5a55d76a96089742ce67ecfea&vid=b46aa5d466a159b5a55d76a96089742ce67ecfea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR