2,2,4-Trimethyl-1,3-pentanediol diisobutyrate ≥98.5%
Supplier: TCI America
Synonyms:
2,2,4-Trimethylpentane-1,3-diyl bis(2-methylpropanoate)
T0997-500ML
T0997-25ML
TCT0997-500MLEA
37.92
CAD
TCT0997-500ML
TCT0997-025ML
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate ≥98.5%
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate
CAS Number: 6846-50-0
MDL Number: MFCD00059267
Molecular Formula: C16H30O4
Molecular Weight: 286.41
Purity/Analysis Method: >98.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 280
Melting point (°C): -70
Flash Point (°C): 121
Specific Gravity (20/20): 0.95
Formula:
C₁₆H₃₀O₄ Boiling Pt: 280 °C Melting Pt: –70 °C Density: 0.95 (20 °C) Flash Pt: 121 °C |
MDL Number:
MFCD00059267 CAS Number: 6846-50-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...