Tris(4-methylphenyl)phosphine oxide ≥98.0%
Supplier: TCI America
Synonyms:
Tri-p-tolylphosphine oxide
T0995-5G
T0995-1G
TCT0995-5GEA
1069.66
CAD
TCT0995-5G
TCT0995-001G
Tris(4-methylphenyl)phosphine oxide ≥98.0%
Tris(4-methylphenyl)phosphine oxide
CAS Number: 797-70-6
MDL Number: MFCD00059321
Molecular Formula: C21H21OP
Molecular Weight: 320.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 145
Other Product Names / Synonyms: Tri-p-tolylphosphine Oxide
Formula:
C₂₁H₂₁OP MW: 320.3646 g/mol Melting Pt: 145 °C |
MDL Number:
MFCD00059321 CAS Number: 797-70-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...