(±)-3,7-Dimethyl-3-octanol ≥98.0% for synthesis

Supplier: TCI America

Synonyms: Tetrahydrolinalool, 3,7-Dimethyloctan-3-ol, 3,7-Dimethyl-3-octanol

T0991-500ML T0991-25ML
TCT0991-500MLEA 186.52 CAD
TCT0991-500ML TCT0991-025ML
(±)-3,7-Dimethyl-3-octanol ≥98.0% for synthesis
(±)-3,7-Dimethyl-3-octanol

CAS Number: 78-69-3
MDL Number: MFCD00004482
Molecular Formula: C10H22O
Molecular Weight: 158.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 196
Flash Point (°C): 84
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: 3,7-Dimethyl-3-octanol

Formula: C₁₀H₂₂O
MW: 158.28 g/mol
Boiling Pt: 196 °C
Density: 0.83 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00004482
CAS Number: 78-69-3
EINECS: 201-133-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c765110eda67c5ab0a63c14ad49caa45bd1f6e9&vid=2c765110eda67c5ab0a63c14ad49caa45bd1f6e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR