Thujone (α and β mixture) ≥70.0%

Supplier: TCI America

T0989-5ML T0989-25ML
TCT0989-005MLEA 121.9 CAD
TCT0989-005ML TCT0989-25ML
Thujone (α and β mixture) ≥70.0%
Thujone (alpha and beta mixture)

CAS Number: 1125-12-8
MDL Number: MFCD00135597
Molecular Formula: C10H16O
Molecular Weight: 152.24
Purity/Analysis Method: >70.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 64
Specific Gravity (20/20): 0.92
Specific rotation [a]20/D: -15 deg (neat)


Product Categories: Bioscience, Bicyclic Monoterpenes
Other Product Names / Synonyms: 1-Isopropyl-4-methylbicyclo[3.1.0]hexan-3-one

Formula: C₁₀H₁₆O
Density: 0.92 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00135597
CAS Number: 1125-12-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3da5e66366f5a31b40aa3321485adec6d7d9383&vid=d3da5e66366f5a31b40aa3321485adec6d7d9383&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 70.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR