N,N',N'',N'''-Tetraacetylglycoluril ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N,N',N'',N'''-Tetraacetylacetyleneurea

T0985-25G T0985-5G
TCT0985-025GEA 454.57 CAD
TCT0985-025G TCT0985-005G
N,N',N'',N'''-Tetraacetylglycoluril ≥98.0% (by HPLC, total nitrogen)
N,N',N'',N'''-Tetraacetylglycoluril

CAS Number: 10543-60-9
MDL Number: MFCD00022618
Molecular Formula: C12H14N4O6
Molecular Weight: 310.27
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 237


Other Product Names / Synonyms: N,N',N'',N'''-Tetraacetylacetyleneurea

Formula: C₁₂H₁₄N₄O₆
MW: 310.27 g/mol
Melting Pt: 237 °C
MDL Number: MFCD00022618
CAS Number: 10543-60-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24669292979d3390a5ba9d2b5e95bb1a1f7f1a56&vid=24669292979d3390a5ba9d2b5e95bb1a1f7f1a56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR