N-(3-Aminopropyl)-2-nitrobenzenesulfonamide hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
N-(2-Nitrobenzenesulphonyl)-1,3-propanediamine Hydrochloride, N-(o-Ns)-1,3-diaminopropane Hydrochloride, 1-Amino-3-(2-nitrobenzenesulphonamido)propane Hydrochloride, 1-Amino-3-(2-nitrobenzenesulfonamido)propane Hydrochloride, N-(2-Nitrobenzenesulfonyl)-1,3-propanediamine Hydrochloride
CAS Number: 863983-46-4
MDL Number: MFCD07366920
Molecular Formula: C9H13N3O4S
Molecular Weight: 295.74
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: 1-Amino-3-(2-nitrobenzenesulfonamido)propane Hydrochloride, N-(2-Nitrobenzenesulfonyl)-1,3-propanediamine Hydrochloride, N-(o-Ns)-1,3-diaminopropane Hydrochloride
Formula:
C₉H₁₃N₃O₄S MW: 295.74 g/mol |
MDL Number:
MFCD07366920 CAS Number: 863983-46-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...