Tetrachloro-o-benzoquinone ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
3,4,5,6-Tetrachloro-1,2-benzoquinone, Tetrachloro-1,2-benzoquinone, 2-Chloranil, Tetrachloro-o-benzoquinone, o-Chloranil, 3,4,5,6-Tetrachlorocyclohexa-3,5-diene-1,2-dione
CAS Number: 2435-53-2
MDL Number: MFCD00001646
Molecular Formula: C6Cl4O2
Molecular Weight: 245.86
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Color: Yellow Red
Melting point (°C): 128
Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: Tetrachloro-o-benzoquinone
Formula:
C₆Cl₄O₂ MW: 245.88 g/mol Boiling Pt: 312 °C (1013 hPa) Melting Pt: 128 °C Density: 2.01 g/cm³ (20 °C) |
MDL Number:
MFCD00001646 CAS Number: 2435-53-2 EINECS: 219-424-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...