Ethyl tetrahydrofuran-2-acetate ≥98.0%

Supplier: TCI America

T0957-25ML T0957-5ML
TCT0957-025MLEA 1164.46 CAD
TCT0957-025ML TCT0957-005ML
Ethyl tetrahydrofuran-2-acetate ≥98.0%
Ethyl tetrahydrofuran-2-acetate

CAS Number: 2434-02-8
MDL Number: MFCD00059735
Molecular Formula: C8H14O3
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 98
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Tetrahydrofuran-2-acetic Acid Ethyl Ester

Formula: C₈H₁₄O₃
Boiling Pt: 98 °C
Density: 1.03 (20 °C)
MDL Number: MFCD00059735
CAS Number: 2434-02-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caaf7d5275ef345ec5b26c1dc98cdd5651707801&vid=caaf7d5275ef345ec5b26c1dc98cdd5651707801&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR