L-Theanine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
T0954-1G
T0954-5G
TCT0954-001GEA
182.81
CAD
TCT0954-001G
TCT0954-005G
L-Theanine ≥98.0% (by titrimetric analysis)
L-Theanine
CAS Number: 3081-61-6
MDL Number: MFCD00059653
Molecular Formula: C7H14N2O3
Molecular Weight: 174.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 207
Specific rotation [a]20/D: 8 deg (C=5, H2O)
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Others (Phytochemicals), non-Proteinogenic Amino Acids
Other Product Names / Synonyms: N-gamma-Ethyl-L-glutamine, L-gamma-Glutamylethylamide
Formula:
C₇H₁₄N₂O₃ Melting Pt: 207 °C |
MDL Number:
MFCD00059653 CAS Number: 3081-61-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...