Triethylene glycol dimethacrylate ≥95.0% stabilized

Supplier: TCI America

Synonyms: (Ethane-1,2-diylbis(oxy))bis(ethane-2,1-diyl) bis(2-methylacrylate)

T0948-500ML T0948-25ML
TCT0948-500MLEA 135.88 CAD
TCT0948-500ML TCT0948-025ML
Triethylene glycol dimethacrylate ≥95.0% stabilized
Triethylene glycol dimethacrylate

(stabilized with MEHQ)
CAS Number: 109-16-0
MDL Number: MFCD00008591
Molecular Formula: C14H22O6
Molecular Weight: 286.32
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 155
Flash Point (°C): 159
Specific Gravity (20/20): 1.07

Formula: C₁₄H₂₂O₆
MW: 286.32 g/mol
Boiling Pt: 155 °C
Density: 1.07 (20 °C)
Flash Pt: 159 °C
MDL Number: MFCD00008591
CAS Number: 109-16-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a33784ec7531646d54b66dd8c0b7cee882308fa0&vid=a33784ec7531646d54b66dd8c0b7cee882308fa0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR