2,2',5,5'-Tetrachlorobenzidine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
T0933-25G
T0933-500G
TCT0933-025GEA
105.86
CAD
TCT0933-025G
TCT0933-500G
2,2',5,5'-Tetrachlorobenzidine ≥98.0% (by HPLC, total nitrogen)
2,2',5,5'-Tetrachlorobenzidine
CAS Number: 15721-02-5
MDL Number: MFCD00043918
Molecular Formula: C12H8Cl4N2
Molecular Weight: 322.01
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 140
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4,4'-Diamino-2,2',5,5'-tetrachlorobiphenyl
Formula:
C₁₂H₈Cl₄N₂ Melting Pt: 140 °C |
MDL Number:
MFCD00043918 CAS Number: 15721-02-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...