Tetrafluoroterephthalic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,3,5,6-Tetrafluoroterephthalic acid

T0930-1G T0930-5G
TCT0930-001GEA 86.9 CAD
TCT0930-001G TCT0930-5G
Tetrafluoroterephthalic acid ≥97.0% (by titrimetric analysis)
Tetrafluoroterephthalic acid

CAS Number: 652-36-8
MDL Number: MFCD00012301
Molecular Formula: C8H2F4O4
Molecular Weight: 238.09
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White

Formula: C₈H₂F₄O₄
MW: 238.0900 g/mol
MDL Number: MFCD00012301
CAS Number: 652-36-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62329ccac1d6960c79e19466441ac8bdf9aa6162&vid=62329ccac1d6960c79e19466441ac8bdf9aa6162&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR