1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene ≥95.0%

Supplier: TCI America

T0928-25G T0928-5G
TCT0928-025GEA 269.39 CAD
TCT0928-025G TCT0928-5G
1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene ≥95.0%
1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene

CAS Number: 2207-27-4
MDL Number: MFCD00001353
Molecular Formula: C7H6Cl4O2
Molecular Weight: 263.92
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 110
Melting point (°C): 27
Flash Point (°C): 67


Other Product Names / Synonyms: 1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene

Formula: C₇H₆Cl₄O₂
Boiling Pt: 110 °C
Melting Pt: 27 °C
Flash Pt: 67 °C
MDL Number: MFCD00001353
CAS Number: 2207-27-4
EINECS: 218-623-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a9c88849febda18bc78401658c5907fffc508dd&vid=3a9c88849febda18bc78401658c5907fffc508dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR