Tetradecyltrimethylammonium chloride ≥98.0% for synthesis

Supplier: TCI America

Synonyms: TTAC, Tetradonium chloride, N,N,N-Trimethyltetradecan-1-aminium chloride, Trimethyl-tetradecylammonium chloride, Myristyltrimethylammonium chloride, n-Tetradecyltrimethylammonium chloride, (1-Tetradecyl)trimethylammonium chloride

T0926-25G T0926-500G
TCT0926-025GEA 165.6 CAD
TCT0926-025G TCT0926-500G
Tetradecyltrimethylammonium chloride ≥98.0% for synthesis
Tetradecyltrimethylammonium chloride

CAS Number: 4574-04-3
MDL Number: MFCD00059968
Molecular Formula: C17H38ClN
Molecular Weight: 291.95
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Materials Chemistry, Cationic Surfactants, Bioscience, Quaternary Ammonium Salts for Clinical Chemistry
Other Product Names / Synonyms: Myristyltrimethylammonium Chloride, Tetradecyltrimethylammonium Chloride, Trimethylmyristylammonium Chloride, TTAC, MTAC

Formula: C₁₇H₃₈ClN
MW: 291.95 g/mol
MDL Number: MFCD00059968
CAS Number: 4574-04-3
EINECS: 224-958-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78deaebc195e4264c5ee44cb1fdc51bb266f2286&vid=78deaebc195e4264c5ee44cb1fdc51bb266f2286&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR