2,4,6-Tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)mesitylene ≥95.0%

Supplier: TCI America

Synonyms: 1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene, 4,4',4''-((2,4,6-Trimethylbenzene-1,3,5-triyl)tris(methylene))tris(2,6-di-tert-butylphenol)

T0916-25G T0916-500G T0916-100G
TCT0916-025GEA 63.14 CAD
TCT0916-025G TCT0916-500G TCT0916-100G
2,4,6-Tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)mesitylene ≥95.0%
2,4,6-Tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)mesitylene

CAS Number: 1709-70-2
MDL Number: MFCD00026284
Molecular Formula: C54H78O3
Molecular Weight: 775.22
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 243

Formula: C₅₄H₇₈O₃
MW: 775.22 g/mol
Melting Pt: 243 °C
MDL Number: MFCD00026284
CAS Number: 1709-70-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a91ca1f16acee2c7996d0a705fe3ffadd3ee5e4&vid=5a91ca1f16acee2c7996d0a705fe3ffadd3ee5e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR