Trimethylolpropane trimethacrylate ≥90.0% stabilized

Supplier: TCI America

Synonyms: 2-Ethyl-2-hydroxymethyl-1,3-propanediol Trimethacrylate, TMPTMA, 2-Ethyl-2-((methacryloyloxy)methyl)propane-1,3-diyl bis(2-methylacrylate)

T0912-25ML T0912-500ML
TCT0912-025MLEA 37.92 CAD
TCT0912-025ML TCT0912-500ML
Trimethylolpropane trimethacrylate ≥90.0% stabilized
Trimethylolpropane trimethacrylate

(stabilized with MEHQ)
CAS Number: 3290-92-4
MDL Number: MFCD00008588
Molecular Formula: C18H26O6
Molecular Weight: 338.40
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 155
Melting point (°C): -14
Flash Point (°C): 182
Specific Gravity (20/20): 1.06


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2-Ethyl-2-hydroxymethyl-1,3-propanediol Trimethacrylate

Formula: C₁₈H₂₆O₆
MW: 338.40 g/mol
Boiling Pt: 155 °C
Melting Pt: –14 °C
Density: 1.06 (20 °C)
Flash Pt: 182 °C
MDL Number: MFCD00008588
CAS Number: 3290-92-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfef6e70843e84ce6cf7db093a2e60aecf997fb6&vid=cfef6e70843e84ce6cf7db093a2e60aecf997fb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR