Ditetradecyl-3,3'-thiodipropionate ≥94.0%
Supplier: TCI America
Synonyms:
3,3'-Thiodipropionic Acid Ditetradecyl Ester, 3,3'-Thiodipropionic Acid Dimyristyl Ester, Dimyristyl 3,3'-Thiodipropionate, 3,3-Thiodipropionicaciddimyristylester
CAS Number: 16545-54-3
MDL Number: MFCD00048462
Molecular Formula: C34H66O4S
Molecular Weight: 570.96
Purity/Analysis Method: >94.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 52
Other Product Names / Synonyms: Dimyristyl 3,3'-Thiodipropionate, 3,3'-Thiodipropionic Acid Dimyristyl Ester, 3,3'-Thiodipropionic Acid Ditetradecyl Ester
Formula:
C₃₄H₆₆O₄S MW: 570.96 g/mol Melting Pt: 52 °C |
MDL Number:
MFCD00048462 CAS Number: 16545-54-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...