Calcium thioglycolate trihydrate ≥94.0%

Supplier: TCI America

Synonyms: Calcium mercaptoacetate trihydrate, Calcium thioglycolate trihydrate, Calcium bis(mercaptoacetate) trihydrate, Mercaptoacetic acid calcium salt trihydrate

T0902-25G T0902-500G
TCT0902-025GEA 28.44 CAD
TCT0902-025G TCT0902-500G
Calcium thioglycolate trihydrate ≥94.0%
Calcium bis(mercaptoacetate) (Calcium thioglycolate) trihydrate

CAS Number: 5793-98-6
MDL Number: MFCD00043437
Molecular Formula: C2H4O2S
Molecular Weight: 130.17
Purity/Analysis Method: >94.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Thioglycolic Acid Calcium Salt, Mercaptoacetic Acid Calcium Salt

Formula: C₄H₆CaO₄S₂·3H₂O
MW: 276.35 g/mol
MDL Number: MFCD00150707
CAS Number: 65208-41-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79177599b6319a311b6ec7ad312a5376e548ed07&vid=79177599b6319a311b6ec7ad312a5376e548ed07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR