Dimethyl-3,3'-thiodipropionate ≥98.0%
Supplier: TCI America
Synonyms:
Dimethyl 3,3-Thiodipropionate, 3,3'-Thiodipropionic Acid Dimethyl Ester
T0901-500ML
T0901-25ML
TCT0901-500MLEA
184.86
CAD
TCT0901-500ML
TCT0901-025ML
Dimethyl-3,3'-thiodipropionate ≥98.0%
Dimethyl-3,3'-thiodipropionate
CAS Number: 4131-74-2
MDL Number: MFCD00008465
Molecular Formula: C8H14O4S
Molecular Weight: 206.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 164
Flash Point (°C): 93
Specific Gravity (20/20): 1.16
Other Product Names / Synonyms: 3,3'-Thiodipropionic Acid Dimethyl Ester
Formula:
C₈H₁₄O₄S MW: 206.26 g/mol Boiling Pt: 164 °C Density: 1.16 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD00008465 CAS Number: 4131-74-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...