Ponceau S ≥85.0% (by total nitrogen basis) for biochemical research

Supplier: TCI America

A2256-1G A2256-5G
TCA2256-1GEA 26.86 CAD
TCA2256-1G TCA2256-5G
Ponceau S ≥85.0% (by total nitrogen basis) for biochemical research
Ponceau S

[for Biochemical Research]
CAS Number: 6226-79-5
MDL Number: MFCD00003892
Molecular Formula: C22H16N4O13S4
Molecular Weight: 760.55
Purity/Analysis Method: >85.0% (N)
Form: Crystal


Product Categories: Bioscience, Stains & Dyes, Analytical Science, Reagents for Staining and Others (Reagents for Electrophoresis)
Other Product Names / Synonyms: Ponceau S

Formula: C₂₂H₁₂N₄Na₄O₁₃S₄
MDL Number: MFCD00003892
CAS Number: 6226-79-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a74a02861af3a3abfd673c83e336e4e6e16e027&vid=dfed7dcf0d1ace09629901baca6a408c749bf0a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR