1,2,4-Trivinylcyclohexane (mixture of isomers) ≥97.0%

Supplier: TCI America

T0899-25ML T0899-500ML
TCT0899-025MLEA 42.66 CAD
TCT0899-025ML TCT0899-500ML
1,2,4-Trivinylcyclohexane (mixture of isomers) ≥97.0%
1,2,4-Trivinylcyclohexane (mixture of isomers)

CAS Number: 2855-27-8
MDL Number: MFCD00001515
Molecular Formula: C12H18
Molecular Weight: 162.28
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 88
Flash Point (°C): 69
Specific Gravity (20/20): 0.85

Formula: C₁₂H₁₈
Boiling Pt: 88 °C
Density: 0.85 (20 °C)
Flash Pt: 69 °C
MDL Number: MFCD00001515
CAS Number: 2855-27-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cda402e14ea03a9775e2d0f69b69ecbb533dd706&vid=cda402e14ea03a9775e2d0f69b69ecbb533dd706&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR