n-Octyl-p-toluenesulfonate ≥96.0% (by GC, titration analysis)

Supplier: TCI America

T0885-500G T0885-25G
TCT0885-500GEA 1312.35 CAD
TCT0885-500G TCT0885-025G
n-Octyl-p-toluenesulfonate ≥96.0% (by GC, titration analysis)
n-Octyl-p-toluenesulfonate

CAS Number: 3386-35-4
MDL Number: MFCD00059447
Molecular Formula: C15H24O3S
Molecular Weight: 284.41
Purity/Analysis Method: >96.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.06


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: p-Toluenesulfonic Acid n-Octyl Ester

Formula: C₁₅H₂₄O₃S
Density: 1.06 (20 °C)
MDL Number: MFCD00059447
CAS Number: 3386-35-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f2dd3b7a253802aeccf4082d165aa289d05aeab&vid=7f2dd3b7a253802aeccf4082d165aa289d05aeab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR