Hexyl-p-toluenesulfonate ≥98.0%

Supplier: TCI America

T0883-25G T0883-500G
TCT0883-025GEA 115.34 CAD
TCT0883-025G TCT0883-500G
Hexyl-p-toluenesulfonate ≥98.0%
Hexyl-p-toluenesulfonate

CAS Number: 3839-35-8
MDL Number: MFCD00059439
Molecular Formula: C13H20O3S
Molecular Weight: 256.36
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 197
Specific Gravity (20/20): 1.09


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: p-Toluenesulfonic Acid Hexyl Ester

Formula: C₁₃H₂₀O₃S
Boiling Pt: 197 °C
Density: 1.09 (20 °C)
MDL Number: MFCD00059439
CAS Number: 3839-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e942f583ef7322422dae697c1a4004a0b36a5468&vid=e942f583ef7322422dae697c1a4004a0b36a5468&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR