Tetrahydrofurfuryl butyrate ≥98.0%

Supplier: TCI America

T0878-25ML T0878-500ML
TCT0878-025MLEA 45.82 CAD
TCT0878-025ML TCT0878-500ML
Tetrahydrofurfuryl butyrate ≥98.0%
Tetrahydrofurfuryl butyrate

CAS Number: 2217-33-6
MDL Number: MFCD00059734
Molecular Formula: C9H16O3
Molecular Weight: 172.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 227
Flash Point (°C): 99
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Butyric Acid Tetrahydrofurfuryl Ester, 2-(Butyryloxymethyl)tetrahydrofuran

Formula: C₉H₁₆O₃
Boiling Pt: 227 °C
Density: 1.01 (20 °C)
Flash Pt: 99 °C
MDL Number: MFCD00059734
CAS Number: 2217-33-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0dd650ec4d4c3cd64f81b4d52f1bf3e2207fe24c&vid=0dd650ec4d4c3cd64f81b4d52f1bf3e2207fe24c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR