(S)-2-Aminobutanamide hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: H-Abu-NH2 · HCl, (2S)-2-Aminobutanamide hydrochloride, (S)-(+)-2-Aminobutanamide hydrochloride, H-Abu-NH2 HCl

A2252-25G A2252-5G
TCA2252-25GEA 205.4 CAD
TCA2252-25G TCA2252-5G
(S)-2-Aminobutanamide hydrochloride ≥98.0% (by HPLC, total nitrogen)
(S)-2-Aminobutanamide hydrochloride

CAS Number: 7682-20-4
MDL Number: MFCD00136565
Molecular Formula: C4H10N2O
Molecular Weight: 138.60
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 263
Specific rotation [a]20/D: 24 deg (C=1, H2O)

Formula: C₄H₁₁ClN₂O
MW: 138.60 g/mol
Melting Pt: 263 °C
MDL Number: MFCD00136565
CAS Number: 7682-20-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7189d549a1fc0b2b9a8214c4d3e2a195e5713df9&vid=7189d549a1fc0b2b9a8214c4d3e2a195e5713df9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR