Tetrabromobisphenol A bis(2-hydroxyethyl)ether ≥93.0%
Supplier: TCI America
Synonyms:
2,2-Bis[3,5-dibromo-4-(2-hydroxyethoxy)phenyl]propane, O,O-Bis(2-hydroxyethyl)tetrabromobisphenol A, 4,4-Isopropylidenebis[2-(2,6-dibromophenoxy)ethanol]
CAS Number: 4162-45-2
MDL Number: MFCD00059603
Molecular Formula: C19H20Br4O4
Molecular Weight: 631.98
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 119
Product Categories: Materials Chemistry, Resin Additives, Monomers, Bisphenol A type Compounds (for High-Performance Polymer Research)
Other Product Names / Synonyms: 2,2-Bis[3,5-dibromo-4-(2-hydroxyethoxy)phenyl]propane, O,O-Bis(2-hydroxyethyl)tetrabromobisphenol A
Formula:
C₁₉H₂₀Br₄O₄ MW: 631.98 g/mol Melting Pt: 119 °C |
MDL Number:
MFCD00059603 CAS Number: 4162-45-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...