Tricyclo[5.2.1.0(2,6)]decanedimethanol ≥90.0%

Supplier: TCI America

Synonyms: Bis(hydroxymethyl)tricyclo[5.2.1.0(2,6)]decane, Tricyclodecanedimethanol (so called)

T0850-500G T0850-25G
TCT0850-500GEA 290.06 CAD
TCT0850-500G TCT0850-025G
Tricyclo[5.2.1.0(2,6)]decanedimethanol ≥90.0%
Tricyclo[5.2.1.0(2,6)]decanedimethanol

CAS Number: 26896-48-0
MDL Number: MFCD00151166
Molecular Formula: C12H20O2
Molecular Weight: 196.29
Purity/Analysis Method: >90.0% (GC)
Form: Starch Syrup
Color: Colorless
Boiling point (°C): 175
Flash Point (°C): 185


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Bis(hydroxymethyl)tricyclo[5.2.1.0(2,6)]decane, Tricyclodecanedimethanol (so called)

Formula: C₁₂H₂₀O₂
MW: 196.29 g/mol
Boiling Pt: 175 °C
Flash Pt: 185 °C
MDL Number: MFCD00151166
CAS Number: 26896-48-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d4c112b018dd0ae244230936015c95c47ea40cf&vid=2d4c112b018dd0ae244230936015c95c47ea40cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR