D-(+)-Trehalose, anhydrous ≥98.0% for synthesis
Supplier: TCI America
Synonyms:
(+)-Trehalose, α,α-Trehalose, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol), Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside, D-(+)-Trehalose Anhydrous, D-Trehalose
T0832-25G
T0832-5G
TCT0832-025GEA
212.29
CAD
TCT0832-025G
TCT0832-5G
D-(+)-Trehalose, anhydrous ≥98.0% for synthesis
D(+)-Trehalose
CAS Number: 99-20-7
MDL Number: MFCD00006628
Molecular Formula: C12H22O11
Molecular Weight: 342.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 205
Specific rotation [a]20/D: 197 deg (C=7, H2O)
Formula:
C₁₂H₂₂O₁₁ MW: 342.3 g/mol Melting Pt: 203 Density: 1.53 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006628 CAS Number: 99-20-7 EINECS: 202-739-6 Merck Index: 12,09713 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...