Tetramethylhydroquinone ≥95.0%

Supplier: TCI America

T0822-1G T0822-5G
TCT0822-001GEA 455.04 CAD
TCT0822-001G TCT0822-5G
Tetramethylhydroquinone ≥95.0%
Tetramethylhydroquinone

CAS Number: 527-18-4
MDL Number: MFCD00045781
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 235


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Dihydroxydurene, Durohydroquinone, 1,4-Dihydroxy-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-1,4-benzenediol

Formula: C₁₀H₁₄O₂
Melting Pt: 235 °C
MDL Number: MFCD00045781
CAS Number: 527-18-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08beed21595c8cd8aa31794b6d39cc1bcb5a9549&vid=08beed21595c8cd8aa31794b6d39cc1bcb5a9549&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR