Tetradecanophenone ≥98.0%

Supplier: TCI America

T0804-25G T0804-5G
TCT0804-025GEA 363.4 CAD
TCT0804-025G TCT0804-5G
Tetradecanophenone ≥98.0%
Tetradecanophenone

CAS Number: 4497-05-6
MDL Number: MFCD00008985
Molecular Formula: C20H32O
Molecular Weight: 288.48
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 206
Melting point (°C): 53


Other Product Names / Synonyms: Myristophenone, Phenyl Tridecyl Ketone

Formula: C₂₀H₃₂O
Boiling Pt: 206 °C
Melting Pt: 53 °C
MDL Number: MFCD00008985
CAS Number: 4497-05-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b400a8900a67eee3b18ed0687e0e9255b473e56&vid=1b400a8900a67eee3b18ed0687e0e9255b473e56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR