p-Tolyldiethanolamine ≥98.0%

Supplier: TCI America

T0797-500G T0797-25G T0797-100G
TCT0797-500GEA 344.85 CAD
TCT0797-500G TCT0797-025G TCT0797-100G
p-Tolyldiethanolamine ≥98.0%
p-Tolyldiethanolamine

CAS Number: 3077-12-1
MDL Number: MFCD00020580
Molecular Formula: C11H17NO2
Molecular Weight: 195.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 163
Melting point (°C): 52


Other Product Names / Synonyms: N,N-Bis(2-hydroxyethyl)-p-toluidine, 2,2'-(4-Methylphenylimino)diethanol, 2,2'-(p-Tolylimino)diethanol

Formula: C₁₁H₁₇NO₂
Boiling Pt: 163 °C
Melting Pt: 52 °C
MDL Number: MFCD00020580
CAS Number: 3077-12-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc310ba02050709b4f8194b52272b297baa0ef65&vid=fc310ba02050709b4f8194b52272b297baa0ef65&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR