Thymoquinone ≥98.0%

Supplier: TCI America

T0795-1G T0795-5G
TCT0795-001GEA 77.74 CAD
TCT0795-001G TCT0795-5G
Thymoquinone ≥98.0%
Thymoquinone

CAS Number: 490-91-5
MDL Number: MFCD00001602
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Yellow
Boiling point (°C): 232
Melting point (°C): 46
Flash Point (°C): 104


Other Product Names / Synonyms: 5-Isopropyl-2-methyl-1,4-benzoquinone, p-Mentha-3,6-diene-2,5-dione

Formula: C₁₀H₁₂O₂
Boiling Pt: 232 °C
Melting Pt: 46 °C
Flash Pt: 104 °C
MDL Number: MFCD00001602
CAS Number: 490-91-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=086f4e47d51d1d1993f7ddd4a28dcce107d93222&vid=086f4e47d51d1d1993f7ddd4a28dcce107d93222&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR