meso-1,2,3,4-Tetrabromobutane ≥98.0%

Supplier: TCI America

T0775-500G T0775-25G
TCT0775-500GEA 696.78 CAD
TCT0775-500G TCT0775-025G
meso-1,2,3,4-Tetrabromobutane ≥98.0%
meso-1,2,3,4-Tetrabromobutane

CAS Number: 2657-67-2
MDL Number: MFCD00070471
Molecular Formula: C4H6Br4
Molecular Weight: 373.71
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 181
Melting point (°C): 116

Formula: BrH₂C(BrCH)₂CH₂Br
Boiling Pt: 181 °C
Melting Pt: 116 °C
MDL Number: MFCD00070471
CAS Number: 2657-67-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=412831859fb4c2fc34d3c9b666d7649493a1cff8&vid=412831859fb4c2fc34d3c9b666d7649493a1cff8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR