2-Tridecenoic acid ≥80.0%
Supplier: TCI America
T0759-5G
T0759-25G
TCT0759-005GEA
77.42
CAD
TCT0759-005G
TCT0759-025G
2-Tridecenoic acid ≥80.0%
CAS Number: 6969-16-0
MDL Number: MFCD00020487
Molecular Formula: C13H24O2
Molecular Weight: 212.33
Purity/Analysis Method: >80.0% (GC)
Form: Crystal
Color: White
Freezing point (°C): 31
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...