Butyl-p-toluenesulfonate ≥97.0%
Supplier: TCI America
T0727-500ML
T0727-25ML
TCT0727-500MLEA
562.48
CAD
TCT0727-500ML
TCT0727-025ML
Butyl-p-toluenesulfonate ≥97.0%
Butyl-p-toluenesulfonate
CAS Number: 778-28-9
MDL Number: MFCD00027203
Molecular Formula: C11H16O3S
Molecular Weight: 228.31
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 172
Specific Gravity (20/20): 1.12
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: p-Toluenesulfonic Acid Butyl Ester
Formula:
C₁₁H₁₆O₃S Boiling Pt: 172 °C Density: 1.12 (20 °C) |
MDL Number:
MFCD00027203 CAS Number: 778-28-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...