Propyl-p-toluenesulfonate ≥98.0%

Supplier: TCI America

Synonyms: p-Toluenesulphonic acid Propyl Ester, p-Toluenesulfonic acid Propyl Ester

T0726-25ML T0726-500ML
TCT0726-025MLEA 90.03 CAD
TCT0726-025ML TCT0726-500ML
Propyl-p-toluenesulfonate ≥98.0%
Propyl-p-toluenesulfonate

CAS Number: 599-91-7
MDL Number: MFCD00059244
Molecular Formula: C10H14O3S
Molecular Weight: 214.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 156
Specific Gravity (20/20): 1.15


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: p-Toluenesulfonic Acid Propyl Ester

Formula: C₁₀H₁₄O₃S
MW: 214.28 g/mol
Boiling Pt: 156 °C
Density: 1.15 (20 °C)
MDL Number: MFCD00059244
CAS Number: 599-91-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a0e3ca7afad296523a8f5b45add4b9faf3b91f4&vid=3a0e3ca7afad296523a8f5b45add4b9faf3b91f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR